missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human SDHD GST (N-Term) Protein

Code produit 18694610 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
10 μg
25 μg
Conditionnement:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18694610 25 μg 25µg
18698520 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 18694610 Fournisseur Novus Biologicals™ Code fournisseur H00006392Q0125ug

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Spécification

À utiliser avec (application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formule 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Identification génétique (Entrez) 6392
Poids moléculaire MolecularWeight-theroretical: 37.18 kDa
Quantité 25 μg
Conditions de stockage Store at -80°C. Avoid freeze-thaw cycles.
Alias de gène CBT1, CII-4, CybS, PGL, PGL1, QPs3, SDH4, succinate dehydrogenase [ubiquinone] cytochrome b small subunit, mitochondrial, Succinate dehydrogenase complex subunit D, succinate dehydrogenase complex, subunit D, integral membrane protein, succinate dehydrogenase ubiquinone cytochrome B small subunit, Succinate-ubiquinone oxidoreductase cytochrome b small subunit, Succinate-ubiquinone reductase membrane anchor subunit
Symbole de gène(s) SDHD
Catégorie de recherche Core ESC Like Genes, Stem Cell Markers
Pureté ou qualité >80% by SDS-PAGE and Coomassie blue staining
Protéine SDHD
Afficher plus Afficher moins
Nom du produit
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.