missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SERPINA5 Protein

Code produit. 15993629
Click to view available options
Quantité:
1 mg
10 μg
2 μg
Conditionnement:
10µg
1mg
2µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 15993629

Marque: enQuireBio™ QP134582ug

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

A cDNA sequence encoding the SERPINA5 was constructed and used to recombinantly synthesize the protein.

Specifications

Nom SERPINA5 Protein
Quantité 2 μg
État réglementaire Research Use Only
Concentration en endotoxines Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Type de produit Recombinant Protein
Réactivité croisée Human
Espèces E. coli
Marqueur de protéine His
Séquence MGSSHHHHHH SSGLVPRGSH MHRHHPREMK KRVEDLHVGA TVAPSSRRDF TFDLYRALAS AAPSQNIFFS PVSISMSLAM LSLGAGSSTK MQILEGLGLN LQKSSEKELH RGFQQLLQEL NQPRDGFQLS LGNALFTDLV VDLQDTFVSA MKTLYLADTF PTNFRDSAGA MKQINDYVAK QTKGKIVDLL KNLDSNAVVI MVNYIFFKAK WETSFNHKGT QEQDFYVTSE TVVRVPMMSR EDQYHYLLDR NLSCRVVGVP YQGNATALFI LPSEGKMQQV ENGLSEKTLR KWLKMFKKRQ LELYLPKFSI EGSYQLEKVL PSLGISNVFT SHADLSGISN HSNIQVSEMV HKAVVEVDES GTRAAAATGT IFTFRSARLN SQRLVFNRPF LMFIVDNNIL FLGKVNRP
Tampon The SERPINA5 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer pH 8.0, 1mM DTT and 10% glycerol.
Pureté ou qualité Greater than 90.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.