missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SHH / Sonic hedgehog Protein

Product Code. 15982998
Click to view available options
Quantité:
100 μg
25 μg
250 μg
5 μg
Unit Size:
100µg
250µg
25µg
5µg
This item is not returnable. View return policy

Product Code. 15982998

Brand: enQuireBio™ QP10181ec100ug

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Human SHH / Sonic hedgehog, was expressed in the hosts and tags indicated.

Specifications

Numéro d’adhésion NP_000184.1
Identification génétique (Entrez) 6469
Nom SHH / Sonic hedgehog Protein
Quantité 100 μg
État réglementaire Research Use Only
Concentration en endotoxines Less than 1.0 EU/ug as determined by LAL method.
Symbole de gène(s) SHH
Longueur des protéines Partial (see sequence information for more details).
Activité biologique The ED50 as determined by inducing alkaline phosphatase production of murine C3H/10T1/2 cells is less than 1 ug/ml, corresponding to a specific activity of > 1.0 M 103 IU/mg.
Type de produit Recombinant Protein
Réactivité croisée Human
Espèces E. coli
Marqueur de protéine Untagged
Séquence IVIGPGRGFG KRRHPKKLTP LAYKQFIPNV AEKTLGASGR YEGKISRNSE RFKELTPNYN PDIIFKDEEN TGADRLMTQR CKDKLNALAI SVMNQWPGVK LRVTEGWDED GHHSEESLHY EGRAVDITTS DRDRSKYGML ARLAVEAGFD WVYYESKAHI HCSVKAENSV AAKSGG
Tampon 0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized
Pureté ou qualité Greater than 98% as determined by SDS-PAGE and HPLC.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.