Learn More
Novus Biologicals™ Recombinant Human SLC35B2 GST (N-Term) Protein
Code produit 18654671 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
2 μg
Conditionnement:
2µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18654671 2 μg 2µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18654671 Fournisseur Novus Biologicals™ Code fournisseur H00347734P012ug
Recombinant Protein
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY
Spécification
À utiliser avec (application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formule 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Identification génétique (Entrez) 347734
Poids moléculaire MolecularWeight-theroretical: 73.9 kDa
Quantité 2 μg
Conditions de stockage Store at -80°C. Avoid freeze/thaw cycles.
Alias de gène adenosine 3 '-phospho 5 '-phosphosulfate transporter 1, PAPS transporter 1, PAPST13'-phosphoadenosine 5 '-phosphosulfate transporter, Putative MAPK-activating protein PM15, Putative NF-kappa-B-activating protein 48, SLL, Solute carrier family 35 member B2, solute carrier family 35 member B2 variant 2, solute carrier family 35, member B2, UGTrel4
Symbole de gène(s) SLC35B2
Pureté ou qualité >80% by SDS-PAGE and Coomassie blue staining
Protéine SLC35B2