missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human SorLA His-tag Protein
Click to view available options
Quantité:
50 μg
Conditionnement:
50µg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human SorLA His-tag Protein is coated at 0.250 μg/mL (100 μL/well), it binds to Recombinant Human PDGF-BB (Catalog # 220-BB ). The ED50 for this binding is 2.00-30.0 ng/mL.
Spécification
Spécification
| Numéro d’adhésion | NP_003096.1 |
| À utiliser avec (application) | Bioactivity |
| Formule | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Identification génétique (Entrez) | 6653 |
| Poids moléculaire | MolecularWeight-observed: 270-345 kDa, under reducing conditions.; MolecularWeight-theroretical: 232 kDa |
| Quantité | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human SorLA protein Ser82-Asp2135, with a C-terminal 6-His tag |
| Conditions de stockage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Concentration en endotoxines | <0.10 EU / 1 μg of the protein by the LAL method. |
| Alias de gène | C11orf32, chromosome 11 open reading frame 32, FLJ21930, FLJ39258, gp250, Low-density lipoprotein receptor relative with 11 ligand-binding repeats, LR11, LR11LDLR relative with 11 ligand-binding repeats, LRP9, mosaic protein LR11, SORL1, SorLA, SorLA-1 SORLA, sortilin-related receptor, sortilin-related receptor, L(DLR class) A repeats containing, sortilin-related receptor, L(DLR class) A repeats-containing, Sorting protein-related receptor containing LDLR class A repeats |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu