Learn More
R&D Systems™ Recombinant Human Sortilin Protein
Code produit. 18367767
Click to view available options
Quantité:
50 μg
Conditionnement:
50µg
Code produit. 18367767
Marque: R&D Systems™ 3154ST050
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
The Recombinant Human Sortilin Protein is derived from NS0. The Recombinant Human Sortilin Protein has been validated for the following applications: Binding Activity.
TRUSTED_SUSTAINABILITY
Spécification
Formule Lyophilized from a 0.2μm filtered solution in PBS.
Identification génétique (Entrez) 6272
Poids moléculaire 76.6 kDa
Nom Sortilin
Quantité 50 μg
Source Mouse myeloma cell line,NS0-derived human Sortilin protein Ser78-Asn755,with a C-terminal 6-His tag
Conditions de stockage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Forme structurelle Monomer
Conjugué Unconjugated
Recombinant Recombinant
Pureté ou qualité 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.