missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human sRank Receptor/TNFRSF11A Protein

Code produit 18638525 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
18638525 500 μg 500µg
18605615 1 mg 1mg
18642496 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18638525 Supplier Novus Biologicals™ Supplier No. NBP235272500ug

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Recombinant Protein

sRank Receptor/TNFRSF11A protein is fully biologically active when compared to standard. The ED50 as determined by its ability to inhibit sRANK Ligand induced nuclear factor kappa B(NFkB) in RAW 264.7 cells is less than 50 ng/mL, corresponding to a specific activity of > 2.0 x 10^4 IU/mg in the presence of 15 ng/mL of recombinant sRANK Ligand.
TRUSTED_SUSTAINABILITY

Specifications

Numéro d’adhésion Q9Y6Q6, Q9Y6Q6, Q9Y6Q6
À utiliser avec (application) SDS-PAGE, Bioactivity
Formule Lyophilized from a 0.2 μm filtered concentrated solution in 20 mM Tris-HCl, pH 8.0, 150 mM NaCl.
Identification génétique (Entrez) 8792
Poids moléculaire MolecularWeight-theroretical: 19.1 kDa
Quantité 500 μg
Conditions de stockage Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Concentration en endotoxines Less than 0.1 EU/μg of sRank Receptor/TNFRSF11A as determined by LAL method.
Alias de gène CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TRANCER, tumor necrosis factor receptor superfamily member 11 A, tumor necrosis factor receptor superfamily, member 11 A, activator of NFKB, tumor necrosis factor receptor superfamily, member 11 A, NFKB activator
Symbole de gène(s) TNFRSF11A
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Pureté ou qualité >98 % pure by SDS-PAGE and HPLC
Protéine sRank Receptor/TNFRSF11A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.