Learn More
enQuireBio™ Recombinant Human STC 2 Protein
Code produit. p-7151514
Click to view available options
Quantité:
10 μg
100 μg
2 μg
Conditionnement:
100µg
10µg
2µg
Code produit. 15904149
Marque: enQuireBio™ QP136132ug
A cDNA sequence encoding the STC 2 was constructed and used to recombinantly synthesize the protein.
Spécification
Identification génétique (Entrez) 8614
Nom STC 2 Protein
Quantité 2 μg
État réglementaire Research Use Only
Concentration en endotoxines Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Symbole de gène(s) STC2
Type de produit Recombinant Protein
Réactivité croisée Human
Espèces Human (HEK293)
Marqueur de protéine Untagged
Séquence TDATNPPEGP QDRSSQQKGR LSLQNTAEIQ HCLVNAGDVG CGVFECFENN SCEIRGLHGI CMTFLHNAGKFDAQGKSFIK DALKCKAHAL RHRFGCISRK CPAIREMVSQ LQRECYLKHD LCAAAQENTR VIVEMIHFKDLLLHEPYVDL VNLLLTCGEE VKEAITHSVQ VQCEQNWGSL CSILSFCTSA IQKPPTAPPE RQPQVDRTKLSRAHHGEAGH HLPEPSSRET GRGAKGERGS KSHPNAHARG RVGGLGAQGP SGSSEWEDEQ SEYSDIRRAAADYKDDDDK
Tampon Filtered (0.4microm) and lyophilized in 0.5 mg/ml in 20mM Tris buffer, 20mM NaCl, pH 7.5.
Pureté ou qualité Greater than 90.0% as determined by SDS-PAGE.