missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Steroid 17-alpha-hydroxylase / 17,20 lyase Protein

A DNA sequence encoding the Homo sapiens (Human) Steroid 17-alpha-hydroxylase/17,20 lyase, was expressed in the hosts and tags indicated.

Marque:  enQuireBio™ QP5904-ec-100ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15978559

  • 476.00€ / 100µg
Expédition estimée: 18-07-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

NP_000093.1
Steroid 17-alpha-hydroxylase / 17,20 lyase Protein
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
1586
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CYP17A1
Recombinant Protein
E. coli
MWELVALLLL TLAYLFWPKR RCPGAKYPKS LLSLPLVGSL PFLPRHGHMH NNFFKLQKKY GPIYSVRMGT KTTVIVGHHQ LAKEVLIKKG KDFSGRPQMA TLDIASNNRK GIAFADSGAH WQLHRRLAMA TFALFKDGDQ KLEKIICQEI STLCDMLATH NGQSIDISFP VFVAVTNVIS LICFNTSYKN GDPELNVIQN YNEGIIDNLS KDSLVDLVPW LKIFPNKTLE KLKSHVKIRN DLLNKILENY KEKFRSDSIT NMLDTLMQAK MNSDNGNAGP DQDSELLSDN HILTTIGDIF GAGVETTTSV VKWTLAFLLH NPQVKKKLYE EIDQNVGFSR TPTISDRNRL LLLEATIREV LRLRPVAPML IPHKANVDSS IGEFAVDKGT EVIINLWALH HNEKEWHQPD QFMPERFLNP AGTQLISPSV SYLPFGAGPR SCIGEILARQ ELFLIMAWLL QRFDLEVPDD GQLPSLEGIP KVVFLIDSFK VKIKVRQAWR EAQAEGST
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions