Learn More
enQuireBio™ Recombinant Human Thymosin beta-4 Protein
Click to view available options
Quantité:
100 μg
20 μg
250 μg
Conditionnement:
100µg
20µg
250µg
Spécification
Spécification
| Numéro d’adhésion | NP_066932.1 |
| Identification génétique (Entrez) | 7114 |
| Nom | Thymosin beta-4 Protein |
| Quantité | 20 μg |
| État réglementaire | Research Use Only |
| Concentration en endotoxines | Less than 1.0 EU/ug as determined by LAL method. |
| Symbole de gène(s) | TMSB4X |
| Longueur des protéines | Partial (see sequence information for more details). |
| Activité biologique | The biological activity determined by its ability to produce a protective effect against hydrogen peroxide in primary lung fibroblasts is in a concentration range of 0.5 - 10 ug/ml. |
| Type de produit | Recombinant Protein |
| Afficher plus |