Learn More
enQuireBio™ Recombinant Human TNFRSF17 / BCMA Protein
Click to view available options
Quantité:
100 μg
20 μg
250 μg
5 μg
Conditionnement:
100µg
20µg
250µg
5µg
Spécification
Spécification
| Numéro d’adhésion | NP_001183.2 |
| Identification génétique (Entrez) | 608 |
| Nom | TNFRSF17 / BCMA Protein |
| Quantité | 5 μg |
| État réglementaire | Research Use Only |
| Concentration en endotoxines | Less than 1.0 EU/ug as determined by LAL method. |
| Symbole de gène(s) | TNFRSF17 |
| Longueur des protéines | Partial (see sequence information for more details). |
| Activité biologique | The ED50 as determined by its ability to inhibit APRIL-mediated proliferation of anti-IgM stimulated murine B cells is no less than 40 ng/ml, corresponding to a specific activity of > 2.5 M 104 IU/mg in the presence of 100 ng/ml human APRIL. |
| Type de produit | Recombinant Protein |
| Afficher plus |