Learn More
enQuireBio™ Recombinant Human TPST1 Protein
Code produit. p-7151708
Click to view available options
Quantité:
1 μg
1 mg
20 μg
5 μg
50 μg
Conditionnement:
1mg
1µg
20µg
50µg
5µg
Code produit. 15984739
Marque: enQuireBio™ QP13799HIS1ug
A cDNA sequence encoding the TPST1 was constructed and used to recombinantly synthesize the protein.
Spécification
Nom TPST1 Protein
Quantité 1 μg
État réglementaire Research Use Only
Concentration en endotoxines Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Type de produit Recombinant Protein
Réactivité croisée Human
Espèces E. coli
Marqueur de protéine His
Séquence MGSSHHHHHH SSGLVPRGSH TGSMQHAMEC HHRIEERSQP VKLESTRTTV RTGLDLKANK TFAYHKDMPL IFIGGVPRSG TTLMRAMLDA HPDIRCGEET RVIPRILALK QMWSRSSKEK IRLDEAGVTD EVLDSAMQAF LLEIIVKHGE PAPYLCNKDP FALKSLTYLS RLFPNAKFLL MVRDGRASVH SMISRKVTIA GFDLNSYRDC LTKWNRAIET MYNQCMEVGY KKCMLVHYEQ LVLHPERWMR TLLKFLQIPW NHSVLHHEEM IGKAGGVSLS KVERSTDQVI KPVNVGALSK WVGKIPPDVL QDMAVIAPML AKLGYDPYAN PPNYGKPDPK IIENTRRVYK GEFQLPDFLK EKPQTEQVE
Tampon TPST1 protein solution (1 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol.
Pureté ou qualité Greater than 90.0% as determined by SDS-PAGE.