missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human uPAR Fc Chimera Protein
Click to view available options
Quantité:
100 μg
Conditionnement:
100µg
Description
When Recombinant Human uPAR Fc Chimera (Catalog # 10378-UK) is immobilized at 1 μg/mL (100 μL/well), Recombinant Human u-Plasminogen Activator (uPA)/Urokinase (Catalog # 1310-SE ) binds with an ED50 of 2-16 ng/mL.
Spécification
Spécification
| Numéro d’adhésion | Q03405.1 |
| Formule | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Identification génétique (Entrez) | 5329 |
| Poids moléculaire | Observed: 74-85 kDa, under reducing conditions; Theroretical: 58 kDa |
| Quantité | 100 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human uPAR protein Human uPAR (Leu23-Arg303) Accession # Q03405.1(N-terminus) IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Conditions de stockage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Concentration en endotoxines | <1.0 EU / 1 μg of the protein by the LAL method. |
| Conjugué | Unconjugated |
| Reconstitution | Reconstitute at 500 μg/mL in PBS. |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu