Learn More
enQuireBio™ Recombinant Human URM1 Protein
Code produit. p-7151817
Click to view available options
Quantité:
1 mg
20 μg
5 μg
Conditionnement:
1mg
20µg
5µg
Code produit. 15905089
Marque: enQuireBio™ QP139065ug
A cDNA sequence encoding the URM1 was constructed and used to recombinantly synthesize the protein.
Spécification
Nom URM1 Protein
Quantité 5 μg
État réglementaire Research Use Only
Concentration en endotoxines Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Type de produit Recombinant Protein
Réactivité croisée Human
Espèces E. coli
Marqueur de protéine His
Séquence MGSSHHHHHH SSGLVPRGSH MAAPLSVEVE FGGGAELLFD GIKKHRVTLP GQEEPWDIRN LLIWIKKNLL KERPELFIQG DSVRPGILVL INDADWELLG ELDYQLQDQD SVLFISTLHG G
Tampon The URM1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 1mM DTT.
Pureté ou qualité Greater than 95.0% as determined by SDS-PAGE.