Learn More
enQuireBio™ Recombinant Human VOPP1 Protein
Code produit. 15995179
Click to view available options
Quantité:
1 mg
10 μg
2 μg
Conditionnement:
10µg
1mg
2µg
Code produit. 15995179
Marque: enQuireBio™ QP139392ug
A cDNA sequence encoding the VOPP1 was constructed and used to recombinantly synthesize the protein.
Spécification
Nom VOPP1 Protein
Quantité 2 μg
État réglementaire Research Use Only
Concentration en endotoxines Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Type de produit Recombinant Protein
Réactivité croisée Human
Espèces E. coli
Marqueur de protéine His
Séquence MGSSHHHHHH SSGLVPRGSH MRRRMYPPPL IEEPAFNVSY TRQPPNPGPG AQQPGPPYYT DPGGPGMNPV GNSMAMAFQV PPNSPQGSVA CPPPPAYCNT PPPPYEQVVK AK
Tampon The VOPP1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 30% glycerol and 0.2M NaCl.
Pureté ou qualité Greater than 90.0% as determined by SDS-PAGE.