Learn More
enQuireBio™ Recombinant Human WWC1 Protein
Code produit. p-7151877
Click to view available options
Quantité:
1 mg
20 μg
5 μg
Conditionnement:
1mg
20µg
5µg
Code produit. 15905269
Marque: enQuireBio™ QP139665ug
A cDNA sequence encoding the WWC1 was constructed and used to recombinantly synthesize the protein.
Spécification
Nom WWC1 Protein
Quantité 5 μg
État réglementaire Research Use Only
Concentration en endotoxines Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Type de produit Recombinant Protein
Réactivité croisée Human
Espèces E. coli
Marqueur de protéine His
Séquence MGSSHHHHHH SSGLVPRGSH MGSEAVGATR IQIALKYDEK NKQFAILIIQ LSNLSALLQQ QDQKVNIRVA VLPCSESTTC LFRTRPLDAS DTLVFNEVFW VSMSYPALHQ KTLRVDVCTT DRSHLEECLG GAQISLAEVC RSGERSTRWY NL
Tampon The WWC1 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT.
Pureté ou qualité Greater than 90% as determined by SDS-PAGE.