missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human XIAP (BIR3) Protein
Click to view available options
Quantité:
50 μg
Unit Size:
50µg
Description
The Recombinant Human XIAP (BIR3) Protein is derived from E. coli. The Recombinant Human XIAP (BIR3) Protein has been validated for the following applications: Inhibition Activity.
Specifications
Specifications
| Formule | Supplied as a 0.2μm filtered solution in HEPES, NaCl and DTT. |
| Identification génétique (Entrez) | 331 |
| Poids moléculaire | 13 kDa |
| Nom | XIAP BIR3 Domain |
| Quantité | 50 μg |
| Source | E. coli-derived human XIAP protein Asn252-Thr356,with an N-terminal Met and 6-His tag |
| Conditions de stockage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Conjugué | Unconjugated |
| Recombinant | Recombinant |
| Pureté ou qualité | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction