missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse BCMA/TNFRSF17 Fc Chimera Protein
Click to view available options
Quantité:
50 μg
Conditionnement:
50µg
Description
The Recombinant Mouse BCMA/TNFRSF17 Fc Chimera Protein is derived from NS0. The Recombinant Mouse BCMA/TNFRSF17 Fc Chimera Protein has been validated for the following applications: Bioactivity.
Spécification
Spécification
| Formule | Lyophilized from a 0.2μm filtered solution in MES and NaCl. |
| Identification génétique (Entrez) | 21935 |
| Poids moléculaire | 32 kDa (monomer) |
| Nom | BCMA/TNFRSF17 Fc Chimera |
| Quantité | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse BCMA/TNFRSF17 protein, (N-terminus) Mouse BCMA (Met1-Thr49) Accession &Num; NP_035738 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Conditions de stockage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Forme structurelle | Disulfide-linked homodimer |
| Alias de gène | B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, TNFRSF13A, TNFRSF17, tumor necrosis factor receptor superfamily, member 17 |
| Conjugué | Unconjugated |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu