missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse C5a Protein

Code produit. 15994188
Click to view available options
Quantité:
1 mg
10 μg
2 μg
Conditionnement:
10µg
1mg
2µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 15994188

Marque: enQuireBio™ QP105412ug

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Denne vare kan ikke returneres. Se returpolitik

A cDNA sequence encoding the C5a was constructed and used to recombinantly synthesize the protein.

Tekniske data

Nom C5a Protein
Quantité 2 μg
État réglementaire Research Use Only
Concentration en endotoxines < 1.0 EU per ug protein as determined by the LAL method.
Activité biologique The ED50 of Recombinant Mouse C5a as determined by its ability to induce N-acetyl-b-D-glucosaminidase release from differentiated U937 human histiocytic lymphoma cells was 5-20ng/ml.
Type de produit Recombinant Protein
Réactivité croisée Mouse
Espèces E. coli
Marqueur de protéine Untagged
Séquence MGLWGILCLL IFLDKTWGQE QTYVISAPKI LRVGSSENVV IQVHGYTEAF DATLSLKSYP DKKVTFSSGY VNLSPENKFQ NAALLTLQPN QVPREESPVS HVYLEVVSKH FSKSKKIPIT YNNGILFIHT DKPVYTPDQS VKIRVYSLGD DLKPAKRETV LTFIDPEGSE VDIVEENDYT GIISFPDFKI PSNPKYGVWT IKANYKKDFT TTGTAYFEIK EYVLPRFSVS IELERTFIGY KNFKNFEITV KARYFYNKVV PDAEVYAFFG LREDIKDEEK QMMHKATQAA KLVDGVAQIS FDSETAVKEL SYNSLEDLNN KYLYIAVTVT ESSGGFSEEA EIPGVKYVLS PYTLNLVATP LFVKPGIPFS IKAQVKDSLE QAVGGVPVTL MAQTVDVNQE TSDLETKRSI THDTDGVAVF VLNLPSNVTV LKFEIRTDDP ELPEENQASK EYEAVAYSSL SQSYIYIAWT ENYKPMLVGE YLNIMVTPKS PYIDKITHYN YLILSKGKIV QYGTREKLFS STYQNINIPV TQNMVPSARL LVYYIVTGEQ TAELVADAVW INIEEKCGNQ LQVHLSPDEY VYSPGQTVSL DMVTEADSWV ALSAVDRAVY KVQGNAKRAM QRVFQALDEK SDLGCGAGGG HDNADVFHLA GLTFLTNANA DDSHYRDDSC KEILRSKRNL HLLRQKIEEQ AAKYKHSVPK KCCYDGARVN FYETCEERVA RVTIGPLCIR AFNECCTIAN KIRKESPHKP VQLGRIHIKT LLPVMKADIR SYFPESWLWE IHRVPKRKQL QVTLPDSLTT WEIQGIGISD NGICVADTLK AKVFKEVFLE MNIPYSVVRG EQIQLKGTVY NYMTSGTKFC VKMSAVEGIC TSGSSAASLH TSRPSRCVFQ RIEGSSSHLV TFTLLPLEIG LHSINFSLET SFGKDILVKT LRVVPEGVKR ESYAGVILDP KGIRGIVNRR KEFPYRIPLD LVPKTKVERI LSVKGLLVGE FLSTVLSKEG INILTHLPKG SAEAELMSIA PVFYVFHYLE AGNHWNIFYP DTLSKRQSLE KKIKQGVVSV MSYRNADYSY SMWKGASAST WLTAFALRVL GQVAKYVKQD ENSICNSLLW LVEKCQLENG SFKENSQYLP IKLQGTLPAE AQEKTLYLTA FSVIGIRKAV DICPTMKIHT ALDKADSFLL ENTLPSKSTF TLAIVAYALS LGDRTHPRFR LIVSALRKEA FVKGDPPIYR YWRDTLKRPD SSVPSSGTAG MVETTAYALL ASLKLKDMNY ANPIIKWLSE EQRYGGGFYS TQDTINAIEG LTEYSLLLKQ IHLDMDINVA YKHEGDFHKY KVTEKHFLGR PVEVSLNDDL VVSTGYSSGL ATVYVKTVVH KISVSEEFCS FYLKIDTQDI EASSHFRLSD SGFKRIIACA SYKPSKEEST SGSSHAVMDI SLPTGIGANE EDLRALVEGV DQLLTDYQIK DGHVILQLNS IPSRDFLCVR FRIFELFQVG FLNPATFTVY EYHRPDKQCT MIYSISDTRL QKVCEGAACT CVEADCAQLQ AEVDLAISAD SRKEKACKPE TAYAYKVRIT SATEENVFVK YTATLLVTYK TGEAADENSE VTFIKKMSCT NANLVKGKQY LIMGKEVLQI KHNFSFKYIY PLDSSTWIEY WPTDTTCPSC QAFVENLNNF AEDLFLNSCE
Tampon Mouse C5a was lyophilized from a concentrated (1 mg/ml) solution in 20mM PBS, pH 7.5 and 350mM NaCl.
Pureté ou qualité Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.