missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CRYGD Protein

A cDNA sequence encoding the CRYGD was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11522-5ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15977518

  • 125.00€ / 5µg
Expédition estimée: 10-07-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

CRYGD Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
CRYGD protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl and 10% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMGKITFY EDRGFQGRHY ECSTDHSNLQ PYFSRCNSVR VDSGCWMLYE QPNFTGCQYF LRRGDYPDYQ QWMGFSDSVR SCRLIPHAGS HRIRLYEREE YRGQMIEFTE DCPSLQDRFH FNEIYSLNVL EGCWVLYDMT NYRGRQYLLR PGEYRRYHDWGAMNARVGSL RRVMDFY
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions