missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse GDF7 Protein

Product Code. 15984458
Click to view available options
Quantité:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15984458

Brand: enQuireBio™ QP1062910ug

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
View alternative products

This item is not returnable. View return policy

A cDNA sequence encoding the GDF7 was constructed and used to recombinantly synthesize the protein.

Specifications

Identification génétique (Entrez) 238057
Nom GDF7 Protein
Quantité 10 μg
État réglementaire Research Use Only
Concentration en endotoxines < 1.0 EU per ug protein as determined by the LAL method.
Symbole de gène(s) Gdf7
Activité biologique The ED50 as determined by inducing alkaline phosphatase production of murine ATDC5 cells is less than 0.5microg/ml, corresponding to a specific activity of > 2000 IU/mg.
Type de produit Recombinant Protein
Réactivité croisée Mouse
Espèces E. coli
Marqueur de protéine Untagged
Séquence TALAGTRGAQ GSGGGGGGGG GGGGGGGGGG GGAGRGHGRR GRSRCSRKSL HVDFKELGWD DWIIAPLDYE AYHCEGVCDF PLRSHLEPTN HAIIQTLLNS MAPDAAPASC CVPARLSPIS ILYIDAANNV VYKQYEDMVV EACGCR
Tampon GDF7 protein was lyophilized from a 0.2microm filtered concentrated solution in 30% Acetonitrile and 0.1% TFA.
Pureté ou qualité Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.