Learn More
enQuireBio™ Recombinant Mouse IgG Protein
A cDNA sequence encoding the IgG was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12386-HIS-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
IgG Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
IgG protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
Insect | |
ADPEPRGPTI KPCPPCKCPA PNLLGGPSVF IFPPKIKDVL MISLSPIVTC VVVDVSEDDP DVQISWFVNN VEVHTAQTQT HREDYNSTLR VVSALPIQHQ DWMSGKEFKC KVNNKDLPAP IERTISKPKG SVRAPQVYVL PPPEEEMTKK QVTLTCMVTD FMPEDIYVEW TNNGKTELNY KNTEPVLDSD GSYFMYSKLR VEKKNWVERN SYSCSVVHEG LHNHHTTKSF SRTPGKHHHH HH |