Learn More
enQuireBio™ Recombinant Mouse S100A3 Protein
Code produit. p-7151280
Click to view available options
Quantité:
1 mg
20 μg
5 μg
Conditionnement:
1mg
20µg
5µg
Code produit. 15903429
Marque: enQuireBio™ QP133935ug
A cDNA sequence encoding the S100A3 was constructed and used to recombinantly synthesize the protein.
Spécification
Nom S100A3 Protein
Quantité 5 μg
État réglementaire Research Use Only
Concentration en endotoxines Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Type de produit Recombinant Protein
Réactivité croisée Mouse
Espèces E. coli
Marqueur de protéine His
Séquence MGSSHHHHHH SSGLVPRGSH MGSHMTRPLE QAVAAIVCTF QEYAGRCGDK YKICQSELKE LLQKELPTWT PSEFRECDYN KFMSVLDTNK DCEVDFGEYV RSLASLCLYC HEYFKECPPE PPCPQ
Tampon S100A3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer, pH 8.0, 10% glycerol, 2mM DTT and 150mM NaCl.
Pureté ou qualité Greater than 95.0% as determined by SDS-PAGE.