missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Bet v 1.0101 Protein

A cDNA sequence encoding the Bet v 1.0101 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11155-50ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15986318

  • 870.00€ / 50µg
Expédition estimée: 17-01-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

Bet v 1.0101 Protein
Research Use Only
Recombinant Protein
Baculovirus
MGVFNYETET TSVIPAARLF KAFILDGDNL FPKVAPQAIS SVENIEGNGG PGTIKKISFP EGFPFKYVKD RVDEVDHTNF KYNYSVIEGG PIGDTLEKIS NEIKIVATPD GGSILKISNK YHTKGDHEVK AEQVKASKEM GETLLRAVES YLLAHSDAYN
Greater than 95.0% as determined by SDS-PAGE.
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Bet v 1.0101 protein solution is supplied in 20mM HEPES buffer pH 8.0, 200mM NaCl and 20% glycerol.
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions