Learn More
enQuireBio™ Recombinant other DERA Protein
A cDNA sequence encoding the DERA was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11658-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
51071 | |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
DERA | |
Other | |
His | |
The DERA solution (1 mg/ml) 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 2mM DTT. |
DERA Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MTDLKASSLR ALKLMDLTTL NDDDTDEKVI ALCHQAKTPV GNTAAICIYP RFIPIARKTL KEQGTPEIRI ATVTNFPHGN DDIDIALAET RAAIAYGADE VDVVFPYRAL MAGNEQVGFD LVKACKEACA AANVLLKVII ETGELKDEAL IRKASEISIK AGADFIKTST GKVAVNATPE SARIMMEVIR DMGVEKTVGF KPAGGVRTAE DAQKYLAIAD ELFGADWADA RHYRFGASSL LASLLKALGH GDGKSASSY |