Learn More
enQuireBio™ Recombinant other Placental Lactogen Caprine Protein
Code produit. p-7148638
Click to view available options
Quantité:
1 mg
200 μg
50 μg
Conditionnement:
1mg
200µg
50µg
Code produit. 15905298
Marque: enQuireBio™ QP1082850ug
A cDNA sequence encoding the Placental Lactogen Caprine was constructed and used to recombinantly synthesize the protein.
Spécification
Nom Placental Lactogen Caprine Protein
Quantité 50 μg
État réglementaire Research Use Only
Concentration en endotoxines < 1.0 EU per ug protein as determined by the LAL method.
Activité biologique Placental Lactogen Caprine is biologically active as evidenced by inducing proliferation of Nb2 cells.
Type de produit Recombinant Protein
Réactivité croisée Other
Espèces E. coli
Marqueur de protéine Untagged
Tampon The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.02-0.03% NaHCO3.
Pureté ou qualité Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.