missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IL 1RA Protein

A cDNA sequence encoding the IL 1RA was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12395-20ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15980279

  • 189.00€ / 20µg
Expédition estimée: 18-07-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

60582
Greater than 90.0% as determined by analysis by SDS-PAGE.
Research Use Only
Il1rn
Rat
His
IL 1RA protein solution (1 mg/ml) containing Phosphate Buffer Saline (pH 7.4), 10% glycerol and 1mM DTT.
IL 1RA Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHPAGKRP CKMQAFRIWD TNQKTFYLRN NQLIAGYLQG PNTKLEEKID MVPIDFRNVF LGIHGGKLCL SCVKSGDDTK LQLEEVNITD LNKNKEEDKR FTFIRSETGP TTSFESLACP GWFLCTTLEA DHPVSLTNTP KEPCTVTKFY FQEDQ
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions