missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat LDHA Protein

Code produit. 15980779
Click to view available options
Quantité:
10 μg
100 μg
2 μg
missing translation for 'unitSize'
100µg
10µg
2µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 15980779

missing translation for 'mfr': enQuireBio™ QP125492ug

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

A cDNA sequence encoding the LDHA was constructed and used to recombinantly synthesize the protein.

Spécification

Identification génétique (Entrez) 24533
Nom LDHA Protein
Quantité 2 μg
État réglementaire Research Use Only
Concentration en endotoxines Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Symbole de gène(s) Ldha
Type de produit Recombinant Protein
Réactivité croisée Rat
Espèces Baculovirus
Marqueur de protéine His
Séquence MAALKDQLIV NLLKEEQVPQ NKITVVGVGA VGMACAISIL MKDLADELAL VDVIEDKLKG EMMDLQHGSL FLKTPKIVSS KDYSVTANSK LVIITAGARQ QEGESRLNLV QRNVNIFKFI IPNVVKYSPQ CKLLIVSNPV DILTYVAWKI SGFPKNRVIG SGCNLDSARF RYLMGERLGV HPLSCHGWVL GEHGDSSVPV WSGVNVAGVS LKSLNPQLGT DADKEQWKDV HKQVVDSAYE VIKLKGYTSW AIGLSVADLA ESIMKNLRRV HPISTMIKGL YGIKEDVFLS VPCILGQNGI SDVVKVTLTP DEEARLKKSA DTLWGIQKEL QFLEHHHHHH
Tampon LDHA protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 1mM DTT and 20% glycerol.
Pureté ou qualité Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.