Learn More
R&D Systems™ Recombinant SARS-CoV-2 XBB.1.5 Spike GCN4-IZ His Protein, CF
Click to view available options
:
100 μg
Description
Measured by its binding ability in a functional ELISA with Recombinant Human ACE-2 Fc Chimera (Catalog # 10544-ZN ).
Spezifikation
Spezifikation
| Numéro d’adhésion | YP_009724390.1 |
| À utiliser avec (application) | Bioactivity |
| Formule | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Identification génétique (Entrez) | 43740568 |
| Poids moléculaire | MolecularWeight-observed: 133-167 kDa, under reducing conditions., MolecularWeight-theroretical: 138 kDa |
| Quantité | 100 μg |
| Source | Human embryonic kidney cell, HEK293-derived sars-cov-2 Spike protein SARS-CoV-2 XBB.1.5 Spike (Val16-Lys1211) (Thr19Ile, Leu24del, Pro25del, Pro26del Ala27Ser, Val83Ala, Gly142Asp, Tyr144del, His146Gln, Gln183Glu, Val213Glu, Gly252Val, Gly339His, Arg346Thr, Leu368Ile, Ser371Phe, Ser373Pro, Ser375Phe, Thr376Ala, Asp405Asn, Arg408Ser, Lys417Asn, Asn440Lys, Val445Pro, Gly446Ser, Asn460Lys, Ser477Asn, Thr478Lys, Glu484Ala, Phe486Pro, Phe490Ser, Gln498Arg, Asn501Tyr, Tyr505His, Asp614Gly, His655Tyr, Asn679Lys, Pro681His, Asn764Lys, Asp796Tyr, Gln954His, Asn969Lys) (Arg682Ser, Arg685Ser, Lys986Pro, Val987Pro) (N-terminus) Accession # YP_009724390.1 GCN4-IZ 6-His tag (C-terminus) |
| Conditions de stockage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution. |
| Concentration en endotoxines | <0.10 EU per 1 μg of the protein by the LAL method. |
| Alias de gène | 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S Protein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur