Learn More
Recombinant VHH Alpaca, anti-Human, anti-Rabbit, Alexa Fluor 488, Clone: CTK0101 and CTK0102, Proteintech ChromoTek
Code produit 17353493
Modifier l'affichage
Click to view available options
Quantité:
10 μL
100 μL
Conditionnement:
100µL
10µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
17353493 10 μL 10µL
17363493 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 17353493 Fournisseur Chromotek Code fournisseur SRBAF488110
 Article restreint
En stock
Ajouter au panier
Ajouter au panier
Alpaca Monoclonal Antibody
Nano-Secondary™ alpaca anti-human IgG/anti-rabbit IgG, recombinant VHH is an anti-human IgG and anti-rabbit IgG specific secondary antibody. It consists of of a mixture of 2 Nanobodies that bind to human IgG and rabbit IgG with high affinity & specificity.
Spécification
Antigène Recombinant VHH
Applications Immunofluorescence, Western Blot, Flow Cytometry
Clone CTK0101 and CTK0102
Espèces hôtes Alpaca
Quantité 10 μL
Espèces cibles Human, Rabbit