Learn More
Recombinant VHH Alpaca, anti-Mouse, Alexa Fluor 488, Clone: CTK0105 and CTK0106, Proteintech ChromoTek
Code produit 17353473
Modifier l'affichage
Click to view available options
Quantité:
10 μL
100 μL
Conditionnement:
100µL
10µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
17353473 10 μL 10µL
17363473 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 17353473 Fournisseur Chromotek Code fournisseur SMS2BAF488110
 Article restreint
En stock
Ajouter au panier
Ajouter au panier
Alpaca Monoclonal Antibody
Nano-Secondary™ alpaca anti-mouse IgG2b, Fc-specific recombinant VHH is an anti-mouse IgG subclass specific antibody. This secondary antibody product consists of Nanobodies that bind to mouse IgG2b with high affinity & specificity.
Spécification
Antigène Recombinant VHH
Applications Immunofluorescence, Western Blot
Clone CTK0105 and CTK0106
Espèces hôtes Alpaca
Quantité 10 μL
Espèces cibles Mouse