Learn More
Recombinant VHH Alpaca, anti-Mouse, Unconjugated, Clone: CTK0104, Proteintech ChromoTek
Code produit 17383483
Modifier l'affichage
Click to view available options
Quantité:
500 μg
Conditionnement:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
17383483 500 μg 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 17383483 Fournisseur Chromotek Code fournisseur smsG1Cys22500
 Article restreint
En stock
Alpaca Monoclonal Antibody
500 μg Nano-Secondary™ for mouse IgG1: alpaca monoclonal Nanobody, Fc-specific VHH, for 2x Cys conjugation, clone [CTK0104].
Spécification
Antigène Recombinant VHH
Applications Immunofluorescence, Western Blot, Flow Cytometry
Clone CTK0104
Espèces hôtes Alpaca
Quantité 500 μg
Espèces cibles Mouse