Learn More
R&D Systems™ Recombinant Viral Dengue Virus 1 NS1 Protein CF
R&D Systems™ Recombinant Viral Dengue Virus 1 NS1 Protein CF is a non-structural protein that is a viral glycoprotein.
Marque: R&D Systems™ 9438-DG-100
Description
The Recombinant Viral Dengue Virus 1 NS1 Protein is derived from HEK293.Spécification
| P17763 | |
| Lyophilized from a 0.2μm filtered solution in PBS. | |
| 100μg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
| Recombinant Protein | |
| Reconstitute at 500μg/mL in PBS. |
| Bioactivity not tested | |
| 41kDa | |
| Human embryonic kidney cell,HEK293-derived viral Dengue Virus 1 NS1 protein Asp776-Ala1127,with a C-terminal 7-His tag | |
| <0.10EU per 1μg of the protein by the LAL method. | |
| Unconjugated |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.