Learn More
R&D Systems™ Reconstitution Buffer 4 (BSA/HCl)
Code produit. 30002149
Click to view available options
Quantité:
2 ml
Conditionnement:
1 pièce
Code produit. 30002149
Marque: R&D Systems™ RB04
0.1% Bovine Serum Albumin in 4 mM HCl
This product is intended as a companion product for reconstitution of selected lyophilized antibody or protein to maximize recovery and activity. Refer to the specific antibody or protein product data sheet for the recommended reconstitution concentrations.
TRUSTED_SUSTAINABILITY
Spécification
Quantité 2 ml