Learn More
Tocris Bioscience™ RECTAS
Click to view available options
Quantité:
10 mg
50 mg
Conditionnement:
10mg
50mg
Description
RECTAS (rectifier of aberrant splicing) is a pre-mRNA splicing modulator of the IKBKAP (inhibitor of κ light polypeptide gene enhancer in B-cells, kinase complex associated protein) gene. It corrects aberrant IKBKAP splicing in vitro in cellular models of familial dysautonomia and in vivo in transgenic mice. In combination with Phenylbutyric acid (Cat. No. 2682), RECTAS rescues PARK7 aberrant splicing and neuronal cell loss in a midbrain organoid model of familial Parkinson's disease. Orally bioavailable and brain penetrant.
Spécification
Spécification
| Nom chimique ou matériau | 2-Chloro-N-(2-furanylmethyl)-7 H-purin-6-amine |
| CAS | 101862-47-9 |
| Quantité | 50 mg |
| Cible | DNA, RNA and Protein Synthesis Compounds |
| Formule moléculaire | C10H8ClN5O |
| Pureté | 0.98 |