Learn More
Novus Biologicals™ Red Blood Cell (RBC) Lysis Buffer
Code produit 18729143 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
50 ml
Conditionnement:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18729143 50 ml 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18729143 Fournisseur Novus Biologicals™ Code fournisseur NBP229442
Applications: Functional
The Red Blood Cell (RBC) Lysis Buffer is for optimal lysis of erythrocytes in single-cell suspensions of mouse hematopoietic tissues such as spleen and human peripheral blood. This buffer contains ammonium chloride, which lyses red cells with minimal effect on lymphocytes. Nucleated red blood cells are not effectively lysed with ammonium chloride. RBC lysis is not necessary when working with mouse thymus and lymph node. Buffer: Dilute 10X RBC Lysis Buffer to 1X in deionized water before use (1mL of 10X RBC lysis buffer with 9mL of deionized water). Bring to room temperature prior to use. Reconstitute: Solubilize the peptides prior to use by making 5mM PBS stock solutions
TRUSTED_SUSTAINABILITY
Spécification
Concentration Functional
À utiliser avec (application) Use in functional reported in scientific literature (PMID: 24371146)
Quantité 50 ml