Learn More
Cole-Parmer Reinforced Thermoplastic Elastomer (TPE) Tubing
Tous les produits Cole-Parmer Produits
Click to view available options
Diamètre (impériale) interne:
0,125 po
0,25 po
0,5 po
0,625 po
0,75 po
1 po
3/8 in.
Diamètre (impériale) externe:
0,375 po
0,5 po
0,625 po
0,75 po
1 in
1,125 po
1,375 po
Longueur totale (impériale):
15 pi
25 pi
50 pi
Conditionnement:
15m
4.50m
7.60m
Description
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. E in., cellent for use in peristaltic pump applications. Great alternative to silicone tubing. Does not contain PVC, DEHP, phthalates, or other plasticizers. Ultra-low e in., tractables and leachables. Low gas and o in., ygen permeability. Low protein binding and nonpyrogenic material. Thermally weldable. USP Class VI, USP 661, and RoHS compliant and BPOG tested
Specifications
Specifications
| Certifications/Conformité | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Diamètre (impériale) interne | 0,125 po |
| Matériau | TPE |
| Diamètre (impériale) externe | 0,375 po |
| Longueur totale (impériale) | 25 pi |
| Méthode de stérilisation | E-beam/gamma irradiation between 25 and 45 kGy |
| Couleur | Translucide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction