Learn More
Thermo Scientific™ Remel™ Tryptic Soy Agar
Code produit. 11914172
Click to view available options
Quantité:
500 g
2,5 kg
Conditionnement:
3kg
500g
Code produit. 11914172
Marque: Thermo Scientific™ R455004
Remel Tryptic Soy Agar (TSA Agar) (Dehydrated) is used for the cultivation of a wide variety of nonfastidious microorganisms.
Remel Tryptic Soy Agar (TSA Agar) (Dehydrated) is used for the cultivation of a wide variety of nonfastidious microorganisms.
TRUSTED_SUSTAINABILITY
Spécification
Description Gélose Trypticase soja
Forme Poudre
Type de produit Gélose
Quantité 2,5 kg
Pour un usage en laboratoire uniquement