Learn More
Labconco™ ReVo Canopy Connection Kit
ReVo Canopy Connection Kit is ideal for ducting ReVo Class II, Type A2 Microbiological Safety Cabinets.
Marque: Labconco™ R3889210
Informations supplémentaires : Poids : 3.40000kg
Description
- Canopy Connection Kit with side exhaust for ducting ReVo Class II, Type A2 Microbiological Safety Cabinets.
Spécification
Canopy Connection Kit | |
465 x 680 x 265 mm | |
900/1200 mm Cabinet | |
465 x 680 x 265 mm |