Learn More
STEMCELL Technologies RoboSep™ HLA Chimerism BC CD56 Positive Selection Kit
Code produit. 17167781
Encompass
Click to view available options
Quantité:
30 ml de couche leuco-plaquettaire
Conditionnement:
1 kit
Code produit. 17167781
Marque: STEMCELL Technologies 17875RF
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
  • The EasySep™ HLA Chimerism Buffy Coat CD56 Positive Selection Kit II is designed to isolate CD56+ cells from freshly prepared buffy coat by positive selection
  • Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD56 and dextran-coated magnetic particles
  • The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • The CD56 antigen is expressed on NK cells and NKT cells
  • This product replaces the EasySep™ Human Buffy Coat CD56 Positive Selection Kit (Catalog #18085) and EasySep™ HLA Buffy Coat CD56 Positive Selection Kit (Catalog #18085HLA), for even faster cell isolations
Spécification
Type de cellule NK Cells
Quantité 30 ml de couche leuco-plaquettaire