missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RPS6KA1 (RSK1), His Tag Recombinant Protein

Code produit. 10312223
Click to view available options
Quantité:
1 mg
100 μg
Conditionnement:
100µg
1mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 10312223

Marque: Invitrogen™ PV4049

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

RPS6KA1 (RSK1) is a growth-factor regulated serine/threonine kinase that is involved in the MAPK cascade. It is thought to control cellular proliferation and differentiation through the phosphorylation of transcription factors. RPS6KA1 contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion Q15418
Concentration See Label
À utiliser avec (application) Kinase Assay
Formule 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Identification génétique (Entrez) 6195
Poids moléculaire 88.5 kDa
Nom Human RPS6KA1 (RSK1), His Tag
Quantité 1 mg
État réglementaire RUO - research use only
Alias de gène 90 kDa ribosomal protein S6 kinase 1; dJ590P13.1 (ribosomal protein S6 kinase, 90 kD, polypeptide 1); hu-1; MAP kinase-activated protein kinase 1; MAP kinase-activated protein kinase 1 A; MAPK-activated protein kinase 1; MAPK-activated protein kinase 1 A; MAPKAP kinase 1; MAPKAP kinase 1 A; MAPKAPK-1; mapkapk1a; MAPKAPK-1 A; p90Rsk; P90-RSK; p90-RSK 1; p90Rsk1; p90-Rsk1; p90S6K; pp90RSK1; ribosomal protein S6 kinase 2 alpha; ribosomal protein S6 kinase 2 beta; ribosomal protein S6 kinase a, polypeptide 1; ribosomal protein S6 kinase A1; ribosomal protein S6 kinase A1 L homeolog; ribosomal protein S6 kinase alpha 1; Ribosomal protein S6 kinase alpha-1; Ribosomal protein S6 kinase II alpha; ribosomal protein S6 kinase II beta; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 90 kD, polypeptide 1; Ribosomal protein S6 kinase, 90 kD, 1; ribosomal protein S6 kinase, 90 kDa, polypeptide 1; ribosomal protein S6 kinase, 90 kDa, polypeptide 1 L homeolog; ribosomal protein S6 kinase, polypeptide A1; ribosomal S6 kinase 1; RPS6KA; Rps6ka1; rps6ka1.L; RPS6KA1L; rsk; RSK1; Rsk-1; S6 kinase II beta; S6 protein kinase (Rsk-1); S6K-alpha 1; S6K-alpha-1; S6KII-alpha; S6KII-beta; wu:fc56h07; XELAEV_18012109mg; zgc:152654
Nom usuel RSK1
Famille des kinases RSK (RPS6K) Kinase Family
Conjugué Unconjugated
Espèces Human
Recombinant Recombinant
Marqueur de protéine His-tag
Séquence Full length
Contenu et stockage -80°C, Avoid Freeze/Thaw Cycles
Système d’expression Insect cells
Forme Liquid
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis