Learn More
MEDICOM Safewear PE Sleeves
Code produit 31781428 Tous les produits Medicom Produits
Modifier l'affichage
Click to view available options
Dimensions:
Universal
Conditionnement:
1000 pièces
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Dimensions unitSize
31781428 Universal 1000 pièces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 31781428 Fournisseur MEDICOM Code fournisseur 20.300
Ajouter au panier
Ajouter au panier
Safewear polyethylene sleeves protect forearms from dirt; lightweight, practical and suitable for food contact.
  • The plasticized and elasticated PE sleeves ensure forearm protection against various types of dirt
  • Food contact
Spécification
Dimensions Universal
Type de produit General Purpose Sleeve
Matériau Polyethylene
Couleur Blue
Stérilité Non-sterile
Jetable Single-use
Type de bouchon Elastic Ends
Type de couture Welded
Longueur (impériale) 15.75 in.
Longueur (métrique) 40 cm