Description
- Disposable hygiene labcoats protect the work environment from particles coming from the operator's body (skin flakes, hair, sweat, etc.)
- They are suitable for HACCP procedures
- They help the operator protect themselves from the risk of stains related to their activity
Spécification
Spécification
| Dimensions | Small |
| Sexe | Unisex |
| Matériau | Polypropylene |
| Couleur | White |
| Type de bouchon | Press Studs |
| Type de manchette | Elastic |
| Jetable | Single-use |
| Stérilité | Non-sterile |
| Résistance | None |
| Modèle | Knee-length |
| Afficher plus |