Learn More
Tocris Bioscience™ SAG
Click to view available options
Quantité:
1 mg
10 mg
Conditionnement:
10mg
1mg
Description
SAG is a potent Smoothened (Smo) receptor agonist (Kd = 59 nM); antagonizes Cyclopamine (Cat. No. 1623) action at the Smo receptor. SAG potently activates the Hedgehog signaling pathway in Shh-light 2 cells (EC50 ∼ 3 nM) and nduces pathway activation independently of Ptch proteins. SAG is a putative inhibitor of a cellular component required for Hedgehog signaling and also enhances neuronal differentiation of iPSCs into dopaminergic neurons.
SAG dihydrochloride (Cat. No. 6390) also available.
Spécification
Spécification
| Quantité | 10 mg |
| Type de produit | SAG |
| Pureté | >99% |
| Poids moléculaire | 490.06 |
| Inhibiteurs | SAG |
Usage exclusivement réservé à la recherche