Learn More
Value Plastics Sanitary Fitting Caps, Polypropylene
Tous les produits Cole-Parmer Produits
Click to view available options
Admission:
0,75 po Mini capuchon sanitaire
1,5 po Sanitary Cap
Conditionnement:
100 pièces
Description
Ideal for capping off fluid transfer system. Sanitary fittings provide a secure connection where preventing contamination is crucial. Constructed of ADCF polypropylene
Spécification
Spécification
| Certifications/Conformité | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Quantité d’emballage | 100/Pk. |
| Admission | 0,75 po Mini capuchon sanitaire |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.