Learn More
IBA Lifesciences SARS-CoV-2 Peptide FIAGLIAIV (HLA-A*0201)
Tous les produits IBA Lifesciences Produits
Click to view available options
Quantité:
1 mg
Conditionnement:
1mg
Description
- SARS-CoV-2 peptide FIAGLIAIV (HLA-A*0201) is a single peptide for stimulation of T cells.
- The peptide from SARS-CoV-2 is synthesized as it is presented by the MHC class I HLA-A*0201 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
- The peptides were purified by HPLC using a water and acetonitrile gradient and are endotoxin-free.
Spécification
Spécification
| Pureté | >85% |
| Niveau d’endotoxines | Free |
| Format | Lyophilized Powder |
| Quantité | 1 mg |
| Catégorie de recherche | T cell stimulation |
Usage exclusivement réservé à la recherche.