Learn More
BOLA Scrubber Columns
Code produit. p-200110020
Click to view available options
Capacité (métrique):
500 ml
1 000 ml
Conditionnement:
1 pièce
Code produit. 10014860
Marque: BOLA A11704
Il en reste null
Ajouter au panier
Ajouter au panier
Scrubber Columns FEP/PTFE
  • Tall, slim scrubber column made of FEP.
  • Inlet and outlet tube as well as riser tube are made of FEP (5.6 x 8 mm), bottom and top are made of pure PTFE.
  • The standard PTFE frit has a pore size of approx. 5μm and is screwed on the riser tube with an M8x1 thread.
  • It can be exchanged with the PTFE gas distributor with fine bores (Cat. No. N 1501-16) which needs a lower primary pressure.
  • Transparent
  • Unbreakable
  • Intense mixing of gas due to tall riser tube
  • Frit easily exchangeable
Spécification
Capacité (métrique) 500 ml
Diamètre (métrique) extérieur 54 mm
Hauteur (métrique) 400 mm