missing translation for 'onlineSavingsMsg'
Learn More

Secoisolariciresinol diglucoside, MedChemExpress

Code produit. 16474008
Encompass_Preferred
Click to view available options
Quantité:
5 mg
10 mg
25 mg
50 mg
100 mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 16474008

Marque: Medchem Express HY105008

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

MedChemExpress Secoisolariciresinol diglucoside ((S,S)-SDG), the main lignan in wholegrain flaxseed, is known for its beneficial effects including anti-inflammatory, antioxidant, anti-mutagenic, anti-microbial, anti-obesity, hypolipidemic, and neuroprotective effects.

Secoisolariciresinol diglucoside ((S,S)-SDG), the main lignan in wholegrain flaxseed, is known for its beneficial effects including anti-inflammatory, antioxidant, anti-mutagenic, anti-microbial, anti-obesity, hypolipidemic, and neuroprotective effects.

Spécification

Nom chimique ou matériau Secoisolariciresinol diglucoside
CAS 257930-74-8
Couleur Blanc
Notes de qualité de la pureté Research
Formule moléculaire C32H46O16
Quantité 10 mg
Synonyme (S,S)-SDG (S,S)-LGM2605
Informations sur la solubilité H2O : ≥ 100 mg/mL (145.62 mM)
SMILES COC1=C(O)C=CC(C[C@H](CO[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)CO)[C@](CO[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)CO)([H])CC4=CC(OC)=C(O)C=C4)=C1
Poids moléculaire (g/mol) 686.7
Poids de la formule 686.7
Pourcentage de pureté 99.74%
Qualité Research
Forme physique Solid
À utiliser avec (application) COVID-19-immunoregulation
Afficher plus Afficher moins

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.