missing translation for 'onlineSavingsMsg'
Learn More

SHP099 hydrochloride, MedChemExpress

Code produit. 16445727
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantité:
5 mg
10 mg
50 mg
100 mg
missing translation for 'unitSize'
10mg
This item is not returnable. View return policy

Product Code. 16445727

missing translation for 'mfr': Medchem Express HY100388A

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

MedChemExpress SHP099 hydrochloride is a potent, selective and orally available SHP2 inhibitor with an IC50 of 70 nM.

SHP099 hydrochloride is a potent, selective and orally available SHP2 inhibitor with an IC50 of 70 nM.

Specifications

Nom chimique ou matériau SHP099 hydrochloride
CAS 2200214-93-1
Couleur Orange
Notes de qualité de la pureté Research
Formule moléculaire C16H20Cl3N5
Quantité 5 mg
Informations sur la solubilité DMSO : 4.1 mg/mL (10.55 mM; Need ultrasonic and warming) ∣H2O : ≥ 2.5 mg/mL (6.43 mM)
SMILES NC1=NC(N2CCC(C)(N)CC2)=CN=C1C3=CC=CC(Cl)=C3Cl.[H]Cl
Poids moléculaire (g/mol) 388.72
Poids de la formule 388.72
Pourcentage de pureté 97.83%
Qualité Research
Forme physique Solid
À utiliser avec (application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.