missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SLK, GST Tag Recombinant Protein
Click to view available options
Quantité:
10 μg
Conditionnement:
10µg
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.
SLK acts as a novel effector of cytoskeletal remodeling and apoptosis, and seems to be ubiquitously expressed. SLK mediates apoptosis and actin stress fiber dissolution.
Spécification
Spécification
| Numéro d’adhésion | Q9H2G2 |
| Concentration | See Label |
| À utiliser avec (application) | Kinase Assay |
| Formule | 50mM tris with 0.02% Triton X-100, 2mM DTT, 150mM NaCl, 0.5mM EDTA, 50% glycerol and no preservative; pH 7.5 |
| Identification génétique (Entrez) | 9748 |
| Poids moléculaire | 160.3 kDa |
| Nom | Human SLK, GST Tag |
| Plage de pH | 7.5 |
| Méthode de purification | Purified |
| Quantité | 10 μg |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu